Gene Transcripts
- Gene Symbol
- CALM3
- Entrez Gene ID
- 808
- Gene Name
- calmodulin 3
- Gene Aliases
- HEL-S-72, PHKD, PHKD3
- Chromosome Location
- Chr. 19: 46601255 - 46610782 on Build GRCh38
- UniGene ID
- Hs.515487
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005184.2 |
NP_005175.2
|
6 |
1367 |
|
View Full Product Details