Gene Transcripts

Gene Symbol
ARHGEF9
Entrez Gene ID
23229
Gene Name
Cdc42 guanine nucleotide exchange factor 9
Gene Aliases
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
Chromosome Location
Chr. X: 63634967 - 63786025 on Build GRCh38
UniGene ID
Hs.54697
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173479.1 NP_001166950.1 9 2725
NM_001173480.1 NP_001166951.1 9 3131
NM_015185.2 NP_056000.1 10 3322
XM_005262249.2 XP_005262306.1 10 3384
XM_005262250.3 XP_005262307.1 10 3282
XM_005262251.1 XP_005262308.1 10 2632
XM_005262252.1 XP_005262309.1 11 2651
XM_011530890.1 XP_011529192.1 11 3066
XM_011530892.1 XP_011529194.1 10 2890
XM_017029363.1 XP_016884852.1 11 3411
XM_017029364.1 XP_016884853.1 12 3188
XM_017029365.1 XP_016884854.1 10 3289
XM_017029366.1 XP_016884855.1 11 3070
XM_017029367.1 XP_016884856.1 11 3059
XM_017029368.1 XP_016884857.1 11 3360
XM_017029369.1 XP_016884858.1 9 3252
XM_017029370.1 XP_016884859.1 9 3161
XM_017029371.1 XP_016884860.1 9 3150
XM_017029372.1 XP_016884861.1 9 2500
XM_017029373.1 XP_016884862.1 11 3425
XM_017029374.1 XP_016884863.1 8 2981
XM_017029375.1 XP_016884864.1 9 3046
XM_017029377.1 XP_016884866.1 9 3049
XM_017029379.1 XP_016884868.1 9 2388
XM_017029380.1 XP_016884869.1 8 2869
GenBank mRNA AB007884.1 - 10 3284

View Full Product Details