Gene Transcripts

Gene Symbol
IFNA1
Entrez Gene ID
3439
Gene Name
interferon alpha 1
Gene Aliases
IFL, IFN, IFN-ALPHA, IFN-alphaD, IFNA13, IFNA@
Chromosome Location
Chr. 9: 21440454 - 21441316 on Build GRCh38
UniGene ID
Hs.37026
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024013.2 NP_076918.1 1 832
GenBank mRNA AB578886.1 - 1 845
V00537.1 - 1 834
V00538.1 - 1 832

View Full Product Details