Gene Transcripts

Gene Symbol
CCL23
Entrez Gene ID
6368
Gene Name
C-C motif chemokine ligand 23
Gene Aliases
CK-BETA-8, CKb8, Ckb-8, Ckb-8-1, MIP-3, MIP3, MPIF-1, SCYA23, hmrp-2a
Chromosome Location
Chr. 17: 36013056 - 36018179 on Build GRCh38
UniGene ID
Hs.169191
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005064.5 NP_005055.3 4 525
NM_145898.3 NP_665905.2 4 474
GenBank mRNA U58913.1 - 4 525
U85767.1 - 4 433

View Full Product Details