Gene Transcripts
- Gene Symbol
- RGS4
- Entrez Gene ID
- 5999
- Gene Name
- regulator of G-protein signaling 4
- Gene Aliases
- RGP4, SCZD9
- Chromosome Location
- Chr. 1: 163068606 - 163076802 on Build GRCh38
- UniGene ID
- Hs.386726
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC000737.1 |
- |
|
1821 |
|
- Gene Symbol
- RPS27
- Entrez Gene ID
- 6232
- Gene Name
- ribosomal protein S27
- Gene Aliases
- MPS-1, MPS1, S27
- Chromosome Location
- Chr. 1: 153990763 - 153992155 on Build GRCh38
- UniGene ID
- Hs.546291
- Species
- Homo sapiens
- Gene Symbol
- ZNF320
- Entrez Gene ID
- 162967
- Gene Name
- zinc finger protein 320
- Gene Aliases
- ZFPL
- Chromosome Location
- Chr. 19: 52863790 - 52897695 on Build GRCh38
- UniGene ID
- Hs.369632
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC064408.1 |
- |
|
1453 |
|
View Full Product Details