Gene Transcripts

Gene Symbol
ZNF628
Entrez Gene ID
89887
Gene Name
zinc finger protein 628
Gene Aliases
ZEC, Zfp628
Chromosome Location
Chr. 19: 55476332 - 55484487 on Build GRCh38
UniGene ID
Hs.525209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033113.2 NP_149104.3 3 807
XM_005259371.3 XP_005259428.1 2 452
GenBank mRNA BC089449.1 - 3 807

View Full Product Details