Gene Transcripts
- Gene Symbol
- MAPKBP1
- Entrez Gene ID
- 23005
- Gene Name
- mitogen-activated protein kinase binding protein 1
- Gene Aliases
- JNKBP-1, JNKBP1
- Chromosome Location
- Chr. 15: 41774434 - 41827855 on Build GRCh38
- UniGene ID
- Hs.513661
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128608.1 |
NP_001122080.1
|
17, 18 |
2219 |
|
NM_001265611.1 |
NP_001252540.1
|
16, 17 |
2201 |
|
NM_014994.2 |
NP_055809.2
|
16, 17 |
2201 |
|
NR_049761.1 |
|
15, 16 |
2011 |
|
NR_049762.1 |
|
14, 15 |
1962 |
|
XM_006720438.1 |
XP_006720501.1
|
15, 16 |
2018 |
|
XM_006720439.2 |
XP_006720502.1
|
3, 4 |
293 |
|
XM_011521383.1 |
XP_011519685.1
|
16, 17 |
2036 |
|
XM_011521384.2 |
XP_011519686.1
|
17, 18 |
2183 |
|
XM_017022017.1 |
XP_016877506.1
|
15, 16 |
2312 |
|
GenBank mRNA |
AB011168.2 |
- |
17, 18 |
2129 |
|
AB177850.1 |
- |
8, 9 |
1049 |
|
AK302482.1 |
- |
14, 15 |
1903 |
|
AL833267.1 |
- |
16, 17 |
2121 |
|
BC036660.1 |
- |
15, 16 |
2016 |
5266613 |
BC113983.1 |
- |
16, 17 |
2093 |
40083186 |
BC114493.1 |
- |
17, 18 |
2111 |
40083182 |
BX640787.1 |
- |
13, 14 |
2366 |
|
BX648786.1 |
- |
16, 17 |
2152 |
|
View Full Product Details