Gene Transcripts
- Gene Symbol
- CCL3
- Entrez Gene ID
- 6348
- Gene Name
- C-C motif chemokine ligand 3
- Gene Aliases
- G0S19-1, LD78ALPHA, MIP-1-alpha, MIP1A, SCYA3
- Chromosome Location
- Chr. 17: 36088256 - 36090160 on Build GRCh38
- UniGene ID
- Hs.514107
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002983.2 |
NP_002974.1
|
2, 3 |
298 |
|
GenBank mRNA |
AF043339.1 |
- |
1, 2 |
130 |
|
BC071834.1 |
- |
2, 3 |
270 |
5935287 |
BC171831.1 |
- |
2, 3 |
232 |
9054485 |
BC171891.1 |
- |
2, 3 |
232 |
9054545 |
BX103455.1 |
- |
2, 3 |
298 |
|
D00044.1 |
- |
2, 3 |
279 |
|
M23452.1 |
- |
2, 3 |
279 |
|
M25315.1 |
- |
2, 3 |
279 |
|
X03754.1 |
- |
2, 3 |
279 |
|
View Full Product Details