Gene Transcripts

Gene Symbol
PGAM2
Entrez Gene ID
5224
Gene Name
phosphoglycerate mutase 2
Gene Aliases
GSD10, PGAM-M, PGAMM
Chromosome Location
Chr. 7: 44062727 - 44065587 on Build GRCh38
UniGene ID
Hs.632642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000290.3 NP_000281.2 2 593
GenBank mRNA BC001904.1 - 2 601
BC073741.1 - 2 589 2823398
CT002473.1 - 2 594
M18172.1 - 2 570

View Full Product Details