Gene Transcripts

Gene Symbol
ABI2
Entrez Gene ID
10152
Gene Name
abl-interactor 2
Gene Aliases
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Chromosome Location
Chr. 2: 203327874 - 203432173 on Build GRCh38
UniGene ID
Hs.471156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282925.1 NP_001269854.1 11 1792
NM_001282926.1 NP_001269855.1 10 1774
NM_001282927.1 NP_001269856.1 9 1267
NM_001282932.1 NP_001269861.1 10 1788
NM_005759.5 NP_005750.4 10 1678
XM_005246217.2 XP_005246274.1 12 2244
XM_005246218.2 XP_005246275.1 11 2226
XM_005246221.2 XP_005246278.1 10 2079
XM_005246225.2 XP_005246282.1 9 1992
XM_005246226.2 XP_005246283.1 9 1956
XM_006712177.2 XP_006712240.1 13 2320
XM_006712179.2 XP_006712242.1 11 2097
XM_006712180.2 XP_006712243.1 11 2061
XM_006712181.2 XP_006712244.1 10 1974
XM_006712182.2 XP_006712245.1 10 1971
XM_006712185.1 XP_006712248.1 10 2032
XM_006712186.2 XP_006712249.1 8 1809
XM_011510472.2 XP_011508774.1 11 2094
XM_011510473.2 XP_011508775.1 11 1543
XM_017003118.1 XP_016858607.1 12 2241
XM_017003119.1 XP_016858608.1 12 2238
XM_017003121.1 XP_016858610.1 11 2154
XM_017003122.1 XP_016858611.1 11 2091
XM_017003123.1 XP_016858612.1 10 2076
XM_017003125.1 XP_016858614.1 10 2040
XM_017003126.1 XP_016858615.1 10 2037
XM_017003128.1 XP_016858617.1 12 2233
XM_017003129.1 XP_016858618.1 10 2010
XM_017003130.1 XP_016858619.1 10 2007
XM_017003131.1 XP_016858620.1 9 1989
XM_017003133.1 XP_016858622.1 12 2173
XM_017003135.1 XP_016858624.1 9 1953
XM_017003137.1 XP_016858626.1 12 2134
XM_017003138.1 XP_016858627.1 10 1911
XM_017003139.1 XP_016858628.1 11 2119
XM_017003140.1 XP_016858629.1 11 2116
XM_017003141.1 XP_016858630.1 9 1896
XM_017003142.1 XP_016858631.1 9 1893
XM_017003143.1 XP_016858632.1 11 2083
XM_017003144.1 XP_016858633.1 9 1827
XM_017003145.1 XP_016858634.1 8 1806
GenBank mRNA AF260261.1 - 11 1516
AK294429.1 - 5 844
AK297537.1 - 6 733
AK299092.1 - 9 1010
AK299672.1 - 10 1169
AK299679.1 - 5 710
AK299688.1 - 7 970
AK299750.1 - 5 616
AK299824.1 - 10 1618
AK302853.1 - 10 1714
AK316343.1 - 5 670
BC001439.2 - 10 1427 3138341
BT009920.1 - 10 1402
U23435.1 - 9 1228
U31089.1 - 9 1147
X95632.1 - 10 1502

View Full Product Details