Gene Transcripts

Gene Symbol
TGIF2LY
Entrez Gene ID
90655
Gene Name
TGFB induced factor homeobox 2 like, Y-linked
Gene Aliases
TGIFLY
Chromosome Location
Chr. Y: 3579085 - 3580041 on Build GRCh38
UniGene ID
Hs.112148
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139214.2 NP_631960.1 2 737
GenBank mRNA AF332223.1 - 2 737
AK292609.1 - 2 781

View Full Product Details