Gene Transcripts
- Gene Symbol
- VWA3A
- Entrez Gene ID
- 146177
- Gene Name
- von Willebrand factor A domain containing 3A
- Gene Aliases
- Chromosome Location
- Chr. 16: 22091930 - 22156966 on Build GRCh38
- UniGene ID
- Hs.10697
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173615.4 |
NP_775886.3
|
14, 15 |
1467 |
|
XM_011545742.2 |
XP_011544044.1
|
16, 17 |
1614 |
|
XM_011545744.2 |
XP_011544046.1
|
1, 2 |
243 |
|
XM_011545746.2 |
XP_011544048.1
|
1, 2 |
243 |
|
XM_017022947.1 |
XP_016878436.1
|
17, 18 |
1687 |
|
XM_017022948.1 |
XP_016878437.1
|
1 |
815 |
|
XM_017022949.1 |
XP_016878438.1
|
1 |
815 |
|
GenBank mRNA |
AK098260.1 |
- |
12, 13 |
1424 |
|
AK128606.1 |
- |
3, 4 |
335 |
|
AK307544.1 |
- |
14, 15 |
1463 |
|
BC109294.1 |
- |
2, 3 |
195 |
|
BC109295.1 |
- |
2, 3 |
195 |
|
View Full Product Details