Gene Transcripts
- Gene Symbol
- MST1
- Entrez Gene ID
- 4485
- Gene Name
- macrophage stimulating 1
- Gene Aliases
- D3F15S2, DNF15S2, HGFL, MSP, NF15S2
- Chromosome Location
- Chr. 3: 49683947 - 49689528 on Build GRCh38
- UniGene ID
- Hs.349110
- Species
- Homo sapiens
- Gene Symbol
- MST1P2
- Entrez Gene ID
- 11209
- Gene Name
- macrophage stimulating 1 pseudogene 2
- Gene Aliases
- MSPL-1, MSPL-2, MSPL-3, MSPL1, MSPL2, MSPL3, MSTP1, MSTP2, MSTP3
- Chromosome Location
- Chr. 1: 16645574 - 16650420 on Build GRCh38
- UniGene ID
- Hs.349110
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK298417.1 |
- |
4 |
353 |
|
AK298457.1 |
- |
4 |
359 |
|
DC362930.1 |
- |
4 |
387 |
|
View Full Product Details