Gene Transcripts

Gene Symbol
Tspan13
Entrez Gene ID
366602
Gene Name
tetraspanin 13
Gene Aliases
Tm4sf13
Chromosome Location
Chr. 6: 55497659 - 55524415 on Build Rnor_6.0
UniGene ID
Rn.24284
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013244.1 NP_001013262.1 6 1447
GenBank mRNA BC089906.1 - 6 1447
FQ215999.1 - 6 1461
FQ217050.1 - 6 1462

View Full Product Details