Gene Transcripts

Gene Symbol
Dph6
Entrez Gene ID
362191
Gene Name
diphthamine biosynthesis 6
Gene Aliases
Atpbd4, RGD1310006
Chromosome Location
Chr. 3: 106007721 - 106136662 on Build Rnor_6.0
UniGene ID
Rn.107116
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014181.1 NP_001014203.1 2 130
GenBank mRNA BC087148.1 - 2 130

View Full Product Details