Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012357.1 | NP_001012357.1 | 4 | 817 | |
XM_008768068.1 | XP_008766290.1 | 3 | 823 | ||
GenBank mRNA | AY863011.1 | - | 4 | 817 | |
BC089865.1 | - | 4 | 817 | 7321336 | |
FQ211273.1 | - | 1 | 304 |