Gene Transcripts

Gene Symbol
Olr1481
Entrez Gene ID
363639
Gene Name
olfactory receptor 1481
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.111155
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000527.1 NP_001000527.1 1 566

View Full Product Details