Gene Transcripts
- Gene Symbol
- OST4
- Entrez Gene ID
- 100128731
- Gene Name
- oligosaccharyltransferase complex subunit 4, non-catalytic
- Gene Aliases
- Chromosome Location
- Chr. 2: 27070472 - 27071699 on Build GRCh38
- UniGene ID
- Hs.502948
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA772256.1 |
- |
1 |
154 |
|
- Gene Symbol
- AGBL5
- Entrez Gene ID
- 60509
- Gene Name
- ATP/GTP binding protein like 5
- Gene Aliases
- CCP5, RP75
- Chromosome Location
- Chr. 2: 27051423 - 27070622 on Build GRCh38
- UniGene ID
- Hs.138207
- Species
- Homo sapiens
View Full Product Details