Gene Transcripts
- Gene Symbol
- MFSD1
- Entrez Gene ID
- 64747
- Gene Name
- major facilitator superfamily domain containing 1
- Gene Aliases
- SMAP4
- Chromosome Location
- Chr. 3: 158801870 - 158829719 on Build GRCh38
- UniGene ID
- Hs.58663
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001167903.1 |
NP_001161375.1
|
7, 8 |
860 |
|
NM_001289406.1 |
NP_001276335.1
|
7, 8 |
864 |
|
NM_001289407.1 |
NP_001276336.1
|
8, 9 |
946 |
|
NM_022736.2 |
NP_073573.2
|
8, 9 |
977 |
|
NR_110328.1 |
|
7, 8 |
864 |
|
XM_006713730.2 |
XP_006713793.1
|
7, 8 |
795 |
|
GenBank mRNA |
AB014732.1 |
- |
8, 9 |
809 |
|
AF351617.2 |
- |
8, 9 |
673 |
|
AK024215.1 |
- |
8, 9 |
780 |
|
AK294628.1 |
- |
7, 8 |
737 |
|
AK297034.1 |
- |
|
568 |
|
AK297593.1 |
- |
8, 9 |
977 |
|
AK300497.1 |
- |
7, 8 |
663 |
|
AK301680.1 |
- |
8, 9 |
716 |
|
BC030542.1 |
- |
7, 8 |
733 |
4915325 |
BC042197.1 |
- |
8, 9 |
789 |
5737259 |
View Full Product Details