Gene Transcripts

Gene Symbol
LCORL
Entrez Gene ID
254251
Gene Name
ligand dependent nuclear receptor corepressor like
Gene Aliases
MLR1
Chromosome Location
Chr. 4: 17841199 - 18023287 on Build GRCh38
UniGene ID
Hs.446201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166139.2 NP_001159611.1 7 1679
XM_017007964.1 XP_016863453.1 8 1555
GenBank mRNA AK055258.1 - 1 567

View Full Product Details