Gene Transcripts

Gene Symbol
1110034G24Rik
Entrez Gene ID
73747
Gene Name
RIKEN cDNA 1110034G24 gene
Gene Aliases
Chromosome Location
Chr. 2: 132686875 - 132751055 on Build GRCm38
UniGene ID
Mm.284114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028637.1 NP_082913.1 1 279
GenBank mRNA BC100475.1 - 1 279

View Full Product Details