Gene Transcripts

Gene Symbol
Dusp6
Entrez Gene ID
116663
Gene Name
dual specificity phosphatase 6
Gene Aliases
Mkp3
Chromosome Location
Chr. 7: 41475163 - 41479393 on Build Rnor_6.0
UniGene ID
Rn.4313
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053883.2 NP_446335.1 2 1101
GenBank mRNA AY724528.1 - 2 1102
BC087003.1 - 2 1101 7124105
U42627.1 - 2 1028
X94185.1 - 2 894

View Full Product Details