Gene Transcripts

Gene Symbol
Crtac1
Entrez Gene ID
171438
Gene Name
cartilage acidic protein 1
Gene Aliases
W307
Chromosome Location
Chr. 1: 261527009 - 261669585 on Build Rnor_6.0
UniGene ID
Rn.198566
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134401.2 NP_599228.2 12 1702
XM_006231385.2 XP_006231447.1 12 1764
GenBank mRNA BC089944.1 - 10 1233
U78304.1 - 3 343

View Full Product Details