Gene Transcripts

Gene Symbol
Cd48
Entrez Gene ID
245962
Gene Name
Cd48 molecule
Gene Aliases
SLAMF2
Chromosome Location
Chr. 13: 90116765 - 90141595 on Build Rnor_6.0
UniGene ID
Rn.3705
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139103.1 NP_620803.1 3 511
GenBank mRNA BC060542.1 - 3 548
FQ228817.1 - 3 565
FQ234539.1 - 3 545
X13016.1 - 3 511

View Full Product Details