Gene Transcripts
- Gene Symbol
- RARB
- Entrez Gene ID
- 5915
- Gene Name
- retinoic acid receptor beta
- Gene Aliases
- HAP, MCOPS12, NR1B2, RARbeta1, RRB2
- Chromosome Location
- Chr. 3: 24829323 - 25597932 on Build GRCh38
- UniGene ID
- Hs.543218
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC069735.1 |
- |
1 |
532 |
|
M62303.1 |
- |
1 |
541 |
|
S82362.1 |
- |
1 |
1525 |
|
View Full Product Details