Gene Transcripts
- Gene Symbol
- Oprd1
- Entrez Gene ID
- 18386
- Gene Name
- opioid receptor, delta 1
- Gene Aliases
- DOR, DOR-1, Nbor, mDOR
- Chromosome Location
- Chr. 4: 132110726 - 132144486 on Build GRCm38
- UniGene ID
- Mm.5243
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043873.1 |
- |
2 |
476 |
|
View Full Product Details