Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001276719.1 | NP_001263648.1 | 4 | 2734 | |
NM_021609.4 | NP_067622.2 | 3 | 2622 | ||
GenBank mRNA | AK028281.1 | - | 3 | 2514 | |
AK145960.1 | - | 4 | 2616 | ||
AK147166.1 | - | 4 | 2626 |