Gene Transcripts
- Gene Symbol
- H2afy2
- Entrez Gene ID
- 404634
- Gene Name
- H2A histone family, member Y2
- Gene Aliases
- macroH2A2
- Chromosome Location
- Chr. 10: 61738647 - 61784104 on Build GRCm38
- UniGene ID
- Mm.272870
- Species
- Mus musculus
- Gene Symbol
- H2afy3
- Entrez Gene ID
- 67552
- Gene Name
- H2A histone family, member Y3
- Gene Aliases
- 4933432H23Rik
- Chromosome Location
- Chr. 15: 62217541 - 62219451 on Build GRCm38
- UniGene ID
- Mm.484027
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017022.1 |
- |
1 |
182 |
|
AK017051.1 |
- |
1 |
231 |
|
View Full Product Details