Gene Transcripts

Gene Symbol
Zc3h6
Entrez Gene ID
78751
Gene Name
zinc finger CCCH type containing 6
Gene Aliases
4631426G04Rik, 4833425H18Rik, AU024389, AW060453, Zc3hdc6, mKIAA2035
Chromosome Location
Chr. 2: 128967402 - 129018563 on Build GRCm38
UniGene ID
Mm.26377
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178404.3 NP_848491.2 2 507
GenBank mRNA AK014766.1 - 2 501
AK039171.1 - 2 502
AK050503.1 - 2 508
BC058173.1 - 2 472

View Full Product Details