Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000432.3 | NP_000423.2 | 7 | 782 | |
GenBank mRNA | BC015821.1 | - | 7 | 764 | |
BC031006.1 | - | 7 | 759 | 4292208 | |
BC031008.1 | - | 7 | 761 | 4297454 | |
BX538165.1 | - | 3 | 292 | ||
M22815.1 | - | 8 | 730 | ||
X57542.1 | - | 8 | 730 | ||
X66141.1 | - | 7 | 741 |