Gene Transcripts

Gene Symbol
Dync1li1
Entrez Gene ID
235661
Gene Name
dynein cytoplasmic 1 light intermediate chain 1
Gene Aliases
1110053F02Rik, Dnclic1, LIC-1
Chromosome Location
Chr. 9: 114688831 - 114723788 on Build GRCm38
UniGene ID
Mm.128627
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146229.2 NP_666341.1 3 323
GenBank mRNA AK082950.1 - 3 364
AK159702.1 - 365
AK180961.1 - 1 66
BC023347.1 - 3 323 5040044

View Full Product Details