Gene Transcripts
- Gene Symbol
- Ncoa7
- Entrez Gene ID
- 211329
- Gene Name
- nuclear receptor coactivator 7
- Gene Aliases
- 9030406N13Rik
- Chromosome Location
- Chr. 10: 30645582 - 30805977 on Build GRCm38
- UniGene ID
- Mm.297903
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042369.1 |
- |
5 |
858 |
|
AK165098.1 |
- |
4 |
1261 |
|
View Full Product Details