Gene Transcripts

Gene Symbol
Adgb
Entrez Gene ID
215772
Gene Name
androglobin
Gene Aliases
9130014G24Rik
Chromosome Location
Chr. 10: 10335703 - 10475135 on Build GRCm38
UniGene ID
Mm.246573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127353.2 NP_001120825.2 16 1805
GenBank mRNA AK078911.1 - 7 780
AK134038.1 - 4 210

View Full Product Details