Gene Transcripts
- Gene Symbol
- BCAT2
- Entrez Gene ID
- 587
- Gene Name
- branched chain amino acid transaminase 2
- Gene Aliases
- BCAM, BCATM, BCT2, PP18
- Chromosome Location
- Chr. 19: 48795062 - 48811063 on Build GRCh38
- UniGene ID
- Hs.512670
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U68418.1 |
- |
9, 10 |
1072 |
|
View Full Product Details