Gene Transcripts

Gene Symbol
PLG
Entrez Gene ID
5340
Gene Name
plasminogen
Gene Aliases
Chromosome Location
Chr. 6: 160702193 - 160754054 on Build GRCh38
UniGene ID
Hs.143436
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000301.3 NP_000292.1 6 762
GenBank mRNA AK312966.1 - 6 713
BC060513.1 - 6 688 4609795
M74220.1 - 6 699
X05199.1 - 6 704

View Full Product Details