Gene Transcripts
- Gene Symbol
- TACC2
- Entrez Gene ID
- 10579
- Gene Name
- transforming acidic coiled-coil containing protein 2
- Gene Aliases
- AZU-1, ECTACC
- Chromosome Location
- Chr. 10: 121989174 - 122254545 on Build GRCh38
- UniGene ID
- Hs.501252
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291878.1 |
NP_001278807.1
|
9, 10 |
2654 |
|
NM_006997.3 |
NP_008928.1
|
8, 9 |
2618 |
|
NM_206860.2 |
NP_996742.1
|
8, 9 |
2618 |
|
NM_206861.2 |
NP_996743.1
|
11, 12 |
2683 |
|
NM_206862.3 |
NP_996744.3
|
14, 15 |
8245 |
|
XM_005269388.1 |
XP_005269445.1
|
15, 16 |
8162 |
|
XM_005269389.4 |
XP_005269446.1
|
15, 16 |
8686 |
|
XM_005269390.1 |
XP_005269447.1
|
16, 17 |
8105 |
|
XM_005269391.1 |
XP_005269448.1
|
14, 15 |
8102 |
|
XM_005269392.1 |
XP_005269449.1
|
15, 16 |
8162 |
|
XM_005269393.1 |
XP_005269450.1
|
15, 16 |
8069 |
|
XM_005269394.1 |
XP_005269451.1
|
14, 15 |
8036 |
|
XM_005269395.1 |
XP_005269452.1
|
14, 15 |
8027 |
|
XM_005269397.1 |
XP_005269454.1
|
13, 14 |
7901 |
|
XM_006717548.1 |
XP_006717611.1
|
15, 16 |
8057 |
|
XM_011539126.1 |
XP_011537428.1
|
15, 16 |
8159 |
|
XM_011539128.1 |
XP_011537430.1
|
13, 14 |
2735 |
|
XM_011539129.1 |
XP_011537431.1
|
12, 13 |
2600 |
|
XM_011539130.1 |
XP_011537432.1
|
12, 13 |
2507 |
|
XM_011539131.1 |
XP_011537433.1
|
8, 9 |
2573 |
|
XM_011539132.1 |
XP_011537434.1
|
9, 10 |
2516 |
|
XM_017015484.1 |
XP_016870973.1
|
15, 16 |
8150 |
|
XM_017015485.1 |
XP_016870974.1
|
16, 17 |
8105 |
|
XM_017015486.1 |
XP_016870975.1
|
15, 16 |
8069 |
|
XM_017015487.1 |
XP_016870976.1
|
15, 16 |
8057 |
|
XM_017015488.1 |
XP_016870977.1
|
13, 14 |
7796 |
|
XM_017015489.1 |
XP_016870978.1
|
13, 14 |
7808 |
|
XM_017015491.1 |
XP_016870980.1
|
8, 9 |
2713 |
|
XM_017015492.1 |
XP_016870981.1
|
8, 9 |
2620 |
|
XM_017015493.1 |
XP_016870982.1
|
8, 9 |
2468 |
|
XM_017015496.1 |
XP_016870985.1
|
9, 10 |
2514 |
|
XM_017015497.1 |
XP_016870986.1
|
9, 10 |
2773 |
|
XM_017015498.1 |
XP_016870987.1
|
8, 9 |
2638 |
|
XM_017015499.1 |
XP_016870988.1
|
11, 12 |
2697 |
|
GenBank mRNA |
AF095791.2 |
- |
8, 9 |
2205 |
|
AF528098.1 |
- |
11, 12 |
2664 |
|
AF528099.1 |
- |
14, 15 |
8226 |
|
AL833304.1 |
- |
13, 14 |
7344 |
|
BC039311.1 |
- |
8, 9 |
2618 |
|
BC141837.1 |
- |
10, 11 |
2306 |
|
BC142673.1 |
- |
10, 11 |
2306 |
|
View Full Product Details