Gene Transcripts

Gene Symbol
ACKR3
Entrez Gene ID
57007
Gene Name
atypical chemokine receptor 3
Gene Aliases
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1
Chromosome Location
Chr. 2: 236537131 - 236582358 on Build GRCh38
UniGene ID
Hs.471751
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020311.2 NP_064707.1 2 450
XM_005246097.2 XP_005246154.1 2 346
XM_005246098.3 XP_005246155.1 3 588
XM_017004516.1 XP_016860005.1 2 341
GenBank mRNA AF030297.1 - 2 298
AK291659.1 - 2 362
BC036661.2 - 4 647 5267661
BM925428.1 - 2 394
DQ822477.1 - 1 236
M64749.1 - 1 236
U67784.1 - 1 56

View Full Product Details