Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 12381234-12381740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPARG
Entrez Gene ID
5468
Gene Name
peroxisome proliferator activated receptor gamma
Gene Aliases
CIMT1, GLM1, NR1C3, PPARG1, PPARG2, PPARgamma
UniGene
Hs.162646
Gene Chromosome Location
Chr.3: 12287850-12471054 on Build GRCh38
Transcripts
NM_005037.5 NM_015869.4 NM_138711.3 NM_138712.3 XM_011533841.2 XM_011533842.2 XM_011533843.2 XM_011533844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4548745 c.455G>A p.G152E Chr.3: 12381466-12381466 on Build GRCh38
6366543 c.370A>T p.N124Y Chr.3: 12381381-12381381 on Build GRCh38
7283718 c.323T>C p.V108A Chr.3: 12381334-12381334 on Build GRCh38
4804800 c.452A>G p.Y151C Chr.3: 12381463-12381463 on Build GRCh38
6053189 c.463G>A p.A155T Chr.3: 12381474-12381474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051540997 Chr.3: 12381512-12381512 on Build GRCh38
rs147975759 Chr.3: 12381449-12381449 on Build GRCh38
rs4135261 Chr.3: 12381539-12381539 on Build GRCh38
rs570632298 Chr.3: 12381259-12381259 on Build GRCh38
rs1017007249 Chr.3: 12381664-12381664 on Build GRCh38
rs749662645 Chr.3: 12381392-12381392 on Build GRCh38
rs995577330 Chr.3: 12381639-12381639 on Build GRCh38
rs957685593 Chr.3: 12381707-12381707 on Build GRCh38
rs373443494 Chr.3: 12381276-12381276 on Build GRCh38
rs752761839 Chr.3: 12381498-12381498 on Build GRCh38
rs556030976 Chr.3: 12381692-12381692 on Build GRCh38
rs757788415 Chr.3: 12381516-12381516 on Build GRCh38
rs766703072 Chr.3: 12381353-12381353 on Build GRCh38
rs781368576 Chr.3: 12381373-12381373 on Build GRCh38
rs796683680 Chr.3: 12381464-12381464 on Build GRCh38
rs907458508 Chr.3: 12381571-12381571 on Build GRCh38
rs763367434 Chr.3: 12381683-12381683 on Build GRCh38
rs768241093 Chr.3: 12381523-12381523 on Build GRCh38
rs945126969 Chr.3: 12381713-12381713 on Build GRCh38
rs755081984 Chr.3: 12381360-12381360 on Build GRCh38
rs1009210673 Chr.3: 12381565-12381565 on Build GRCh38
rs377418765 Chr.3: 12381518-12381518 on Build GRCh38
rs747687206 Chr.3: 12381536-12381536 on Build GRCh38
rs183354275 Chr.3: 12381558-12381558 on Build GRCh38
rs1005736782 Chr.3: 12381628-12381628 on Build GRCh38
rs756575717 Chr.3: 12381382-12381382 on Build GRCh38
rs906749154 Chr.3: 12381569-12381569 on Build GRCh38
rs909871980 Chr.3: 12381377-12381377 on Build GRCh38
rs775193198 Chr.3: 12381279-12381279 on Build GRCh38
rs4135324 Chr.3: 12381614-12381614 on Build GRCh38
rs767364009 Chr.3: 12381582-12381582 on Build GRCh38
rs751417943 Chr.3: 12381470-12381470 on Build GRCh38
rs758183823 Chr.3: 12381347-12381347 on Build GRCh38
rs567719997 Chr.3: 12381702-12381702 on Build GRCh38
rs752075145 Chr.3: 12381697-12381697 on Build GRCh38
rs765206131 Chr.3: 12381429-12381429 on Build GRCh38
rs531436395 Chr.3: 12381344-12381344 on Build GRCh38
rs559238043 Chr.3: 12381647-12381647 on Build GRCh38
rs922629558 Chr.3: 12381670-12381670 on Build GRCh38
rs763646361 Chr.3: 12381290-12381290 on Build GRCh38
rs898483180 Chr.3: 12381622-12381622 on Build GRCh38
rs984243899 Chr.3: 12381324-12381324 on Build GRCh38
rs78013505 Chr.3: 12381305-12381305 on Build GRCh38
rs746236883 Chr.3: 12381522-12381522 on Build GRCh38
rs771328613 Chr.3: 12381407-12381407 on Build GRCh38
rs778203686 Chr.3: 12381389-12381389 on Build GRCh38
rs751871408 Chr.3: 12381357-12381357 on Build GRCh38
rs553578064 Chr.3: 12381543-12381543 on Build GRCh38
rs549579646 Chr.3: 12381420-12381420 on Build GRCh38
rs962781345 Chr.3: 12381307-12381307 on Build GRCh38
rs765091444 Chr.3: 12381333-12381333 on Build GRCh38
rs41484149 Chr.3: 12381531-12381531 on Build GRCh38
rs199870649 Chr.3: 12381473-12381473 on Build GRCh38
rs966671737 Chr.3: 12381678-12381678 on Build GRCh38
rs778314547 Chr.3: 12381513-12381513 on Build GRCh38
rs961763180 Chr.3: 12381665-12381665 on Build GRCh38
rs1028136694 Chr.3: 12381380-12381380 on Build GRCh38
rs749811533 Chr.3: 12381514-12381514 on Build GRCh38
rs567927891 Chr.3: 12381712-12381712 on Build GRCh38
rs761770364 Chr.3: 12381314-12381314 on Build GRCh38
rs746616834 Chr.3: 12381410-12381410 on Build GRCh38
rs756216884 Chr.3: 12381509-12381509 on Build GRCh38
rs556795860 Chr.3: 12381691-12381691 on Build GRCh38
rs773089349 Chr.3: 12381430-12381430 on Build GRCh38
rs148037954 Chr.3: 12381361-12381361 on Build GRCh38
rs753368111 Chr.3: 12381299-12381299 on Build GRCh38
rs750711994 Chr.3: 12381497-12381498 on Build GRCh38
rs1042449284 Chr.3: 12381568-12381568 on Build GRCh38
rs754336834 Chr.3: 12381618-12381618 on Build GRCh38
rs1028362568 Chr.3: 12381657-12381657 on Build GRCh38
rs761245370 Chr.3: 12381428-12381428 on Build GRCh38
rs1003830216 Chr.3: 12381594-12381594 on Build GRCh38
rs767853271 Chr.3: 12381484-12381484 on Build GRCh38
rs1800571 Chr.3: 12381349-12381349 on Build GRCh38
rs267599576 Chr.3: 12381409-12381409 on Build GRCh38
rs779221019 Chr.3: 12381498-12381498 on Build GRCh38
rs549324809 Chr.3: 12381676-12381676 on Build GRCh38
rs760150781 Chr.3: 12381285-12381285 on Build GRCh38
rs574162116 Chr.3: 12381689-12381689 on Build GRCh38
rs527860119 Chr.3: 12381669-12381669 on Build GRCh38
rs369293686 Chr.3: 12381350-12381350 on Build GRCh38
rs910369988 Chr.3: 12381687-12381687 on Build GRCh38
rs748205580 Chr.3: 12381381-12381381 on Build GRCh38
rs762777341 Chr.3: 12381432-12381432 on Build GRCh38

View Full Product Details