Gene Transcripts
- Gene Symbol
- Asb8
- Entrez Gene ID
- 78541
- Gene Name
- ankyrin repeat and SOCS box-containing 8
- Gene Aliases
- 4930539L19Rik, AI788835, C430011H06Rik
- Chromosome Location
- Chr. 15: 98134637 - 98165602 on Build GRCm38
- UniGene ID
- Mm.200766
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170710.1 |
NP_001164181.1
|
3, 4 |
377 |
|
NM_001170711.1 |
NP_001164182.1
|
3, 4 |
383 |
|
NM_030121.4 |
NP_084397.3
|
3, 4 |
373 |
|
GenBank mRNA |
AF398969.1 |
- |
3, 4 |
300 |
|
AK016001.1 |
- |
3, 4 |
431 |
|
AK021239.1 |
- |
3, 4 |
314 |
|
AK030168.1 |
- |
3, 4 |
316 |
|
AK087299.1 |
- |
3, 4 |
335 |
|
AK182854.1 |
- |
2, 3 |
133 |
|
AK214224.1 |
- |
2, 3 |
133 |
|
BC025106.1 |
- |
3, 4 |
297 |
4194829 |
View Full Product Details