Gene Transcripts

Gene Symbol
Cntn5
Entrez Gene ID
244682
Gene Name
contactin 5
Gene Aliases
6720426O10Rik, A830025P08Rik, Gm507, NB-2
Chromosome Location
Chr. 9: 9582665 - 10904775 on Build GRCm38
UniGene ID
Mm.187021
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033359.2 NP_001028531.2 6 833
NM_001170787.1 NP_001164258.1 10 1497
GenBank mRNA AK138164.1 - 6 834
AK162856.1 - 6 833

View Full Product Details