Gene Transcripts

Gene Symbol
Cntnap5a
Entrez Gene ID
636808
Gene Name
contactin associated protein-like 5A
Gene Aliases
Caspr5-1, EG636808
Chromosome Location
Chr. 1: 115684497 - 116587323 on Build GRCm38
UniGene ID
Mm.101576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077425.1 NP_001070893.1 15, 16 2354

View Full Product Details