Gene Transcripts

Gene Symbol
Csn1s1
Entrez Gene ID
24284
Gene Name
casein alpha s1
Gene Aliases
Casa, Csn1, Csna
Chromosome Location
Chr. 14: 21993450 - 22009308 on Build Rnor_6.0
UniGene ID
Rn.10744
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138874.2 NP_620229.2 27 817
GenBank mRNA J00710.1 - 27 817

View Full Product Details