Gene Transcripts

Gene Symbol
Cspg5
Entrez Gene ID
50568
Gene Name
chondroitin sulfate proteoglycan 5 (neuroglycan C)
Gene Aliases
Caleb, Ngc
Chromosome Location
Chr. 8: 118333695 - 118348040 on Build Rnor_6.0
UniGene ID
Rn.10146
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019284.1 NP_062157.1 2 786
NM_133652.1 NP_598413.1 2 774
GenBank mRNA AF292102.1 - 2 774
U33553.1 - 2 786

View Full Product Details