Gene Transcripts

Gene Symbol
Adra2c
Entrez Gene ID
24175
Gene Name
adrenoceptor alpha 2C
Gene Aliases
Chromosome Location
Chr. 14: 80730307 - 80732010 on Build Rnor_6.0
UniGene ID
Rn.10297
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138506.1 NP_612515.1 1 1215
GenBank mRNA M58316.1 - 1 1215

View Full Product Details