Gene Transcripts

Gene Symbol
Krt17
Entrez Gene ID
287702
Gene Name
keratin 17, type I
Gene Aliases
Ka17
Chromosome Location
Chr. 10: 88158993 - 88163712 on Build Rnor_6.0
UniGene ID
Rn.106755
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212545.2 NP_997710.1 6, 7 1241
GenBank mRNA BC100058.1 - 6, 7 1241

View Full Product Details