Gene Transcripts

Gene Symbol
Pmpca
Entrez Gene ID
296588
Gene Name
peptidase (mitochondrial processing) alpha
Gene Aliases
Chromosome Location
Chr. 3: 3834262 - 3842059 on Build Rnor_6.0
UniGene ID
Rn.11175
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003673.1 NP_001003673.1 4, 5 442
GenBank mRNA BC079004.1 - 4, 5 442
M57728.1 - 4, 5 436

View Full Product Details