Gene Transcripts

Gene Symbol
Pes1
Entrez Gene ID
289740
Gene Name
pescadillo ribosomal biogenesis factor 1
Gene Aliases
Chromosome Location
Chr. 14: 84211600 - 84227980 on Build Rnor_6.0
UniGene ID
Rn.9521
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044228.1 NP_001037693.1 5 511
GenBank mRNA BC105910.1 - 5 511

View Full Product Details