Gene Transcripts

Gene Symbol
Cntnap5a
Entrez Gene ID
297832
Gene Name
contactin associated protein-like 5A
Gene Aliases
Caspr5-1, RGD1559812
Chromosome Location
Chr. 13: 6510703 - 6608827 on Build Rnor_6.0
UniGene ID
Rn.202802
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047865.1 NP_001041330.1 17

View Full Product Details