Gene Transcripts

Gene Symbol
Rprm
Entrez Gene ID
680110
Gene Name
reprimo, TP53 dependent G2 arrest mediator candidate
Gene Aliases
Chromosome Location
Chr. 3: 39595297 - 39598371 on Build Rnor_6.0
UniGene ID
Rn.45197
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044276.1 NP_001037741.1 1 523
XM_008761775.1 XP_008759997.1 2 2072
GenBank mRNA BC091400.1 - 1 523

View Full Product Details