Gene Transcripts

Gene Symbol
SDK1
Entrez Gene ID
221935
Gene Name
sidekick cell adhesion molecule 1
Gene Aliases
Chromosome Location
Chr. 7: 3301448 - 4269000 on Build GRCh38
UniGene ID
Hs.653013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079653.1 NP_001073121.1 1 112
GenBank mRNA AL833354.1 - 1 109
DB065853.1 - 1 108
DB454012.1 - 1 112

View Full Product Details